Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number FSA148801000
Price (MYR)
626.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
626.00
Add to cart
Chemical Identifiers
CAS59-52-9
IUPAC Name2,3-disulfanylpropan-1-ol
Molecular FormulaC3H8OS2
InChI KeyWQABCVAJNWAXTE-UHFFFAOYNA-N
SMILESOCC(S)CS
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (Iodometric Titration)≥96.0% to ≤104.0%
Refractive Index1.5700-1.5750 @ 20°C