Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number FSA148800250
Price (MYR)
290.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
290.00
Add to cart
Chemical Identifiers
CAS108-20-3
IUPAC Name2-(propan-2-yloxy)propane
Molecular FormulaC6H14O
InChI KeyZAFNJMIOTHYJRJ-UHFFFAOYSA-N
SMILESCC(C)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Refractive index1.3670 to 1.3690 (20°C, 589 nm)
Acidity=<0.005 %
GC>=99.0 %
Residue after evaporation=<0.01 %