4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals
CAS: 586-76-5 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA148701000
Price (MYR)
829.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
829.00
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metal0.002% max.