Methyl dichloroacetate, 99%, Thermo Scientific Chemicals
Methyl dichloroacetate, 99%, Thermo Scientific Chemicals
Methyl dichloroacetate, 99%, Thermo Scientific Chemicals
Methyl dichloroacetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl dichloroacetate, 99%, Thermo Scientific Chemicals
CAS: 116-54-1 | C3H4Cl2O2 | 142.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA148621000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl dichloroacetate
Name Note99+%
CAS116-54-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: