Malonaldehyde bis(dimethyl acetal), 98%, Thermo Scientific Chemicals
Malonaldehyde bis(dimethyl acetal), 98%, Thermo Scientific Chemicals
Malonaldehyde bis(dimethyl acetal), 98%, Thermo Scientific Chemicals
Malonaldehyde bis(dimethyl acetal), 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonaldehyde bis(dimethyl acetal), 98%, Thermo Scientific Chemicals
CAS: 102-52-3 | C7H16O4 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA148615000
Price (MYR)
951.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
951.00
Add to cart
Chemical Identifiers
CAS70523-22-7
IUPAC Name(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)boronic acid
Molecular FormulaC4H5BN2O4
InChI KeyPVEJOCQTIVCDNO-UHFFFAOYSA-N
SMILESOB(O)C1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (HPLC)≥94.0%
Proton NMRConforms to structure