Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number FSA148530250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS80-15-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<450 APHA
Infrared spectrumConforms
Titration Iodometric80 to 85 %