Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
CAS: 98-10-2 | C6H7NO2S | 157.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA148491000
Price (MYR)
257.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
257.00
Add to cart
Chemical Identifiers
CAS617-27-6
IUPAC Nameethyl 2-aminopropanoate hydrochloride
Molecular FormulaC5H12ClNO2
InChI KeyJCXLZWMDXJFOOI-UHFFFAOYNA-N
SMILESCl.CCOC(=O)C(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Appearance (Color)White