Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
CAS: 98-10-2 | C6H7NO2S | 157.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number FSA148490050
Price (MYR)
236.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
236.00
Add to cart
Chemical Identifiers
CAS1871-76-7
IUPAC Name2,2-diphenylacetyl chloride
Molecular FormulaC14H11ClO
InChI KeyMSYLETHDEIJMAF-UHFFFAOYSA-N
SMILESClC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Titration RCOCl96.5 to 103.5 %
Appearance (Form)Crystalline powder or crystals
Melting point51°C to 56°C