Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA148482500
Price (MYR)
332.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
332.00
Add to cart
Chemical Identifiers
CAS603-69-0
IUPAC Nameethyl 2-acetyl-3-oxobutanoate
Molecular FormulaC8H12O4
InChI KeyYMCDYRGMTRCAPZ-UHFFFAOYSA-N
SMILESCCOC(=O)C(C(C)=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.4700-1.4750 @ 20?C
Appearance (Color)Clear colorless to yellow