Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA148480010
Price (MYR)
886.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
886.00
Add to cart
Chemical Identifiers
CAS7778-50-9
IUPAC Namedipotassium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2K2O7
InChI KeyKMUONIBRACKNSN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration Iodometric>=99.0 %
Insoluble matter=<0.005 %
Chloride (Cl)=<10 ppm
Calcium (Ca)=<30 ppm