L-5-Hydroxytryptophan, 99%
L-5-Hydroxytryptophan, 99%
L-5-Hydroxytryptophan, 99%
L-5-Hydroxytryptophan, 99%
Thermo Scientific Chemicals
L-5-Hydroxytryptophan, 99%
C11H12N2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass Bottle
Catalog number FSA148290250
Price (MYR)
2,867.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,867.00
Add to cart
Specifications
Chemical Name or MaterialL-5-Hydroxytryptophan
Health Hazard 1Warning
Arsenic (As)1ppm max.
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.