2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA147975000
Price (MYR)
425.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
425.00
Add to cart
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofATitrable acid: ≤5 mL (≤0.001 meq/g, ≤0.056 mgKOH/g)
Water Content (Karl Fischer Titration)≤0.1%
FormLiquid
Assay from Supplier's CofA≥99.9% (purity by GC)
Appearance (Color)Clear colorless