2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic bottle
Catalog number FSA147970010
Price (MYR)
737.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
737.00
Add to cart
Chemical Identifiers
CAS123-11-5
IUPAC Name4-methoxybenzaldehyde
Molecular FormulaC8H8O2
InChI KeyZRSNZINYAWTAHE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Liquid
Relative density(D20/4°C) 1.120 to 1.124
Appearance (Color)Clear colorless to yellow