Formic acid, 98+%, pure, Thermo Scientific Chemicals
Formic acid, 98+%, pure, Thermo Scientific Chemicals
Formic acid, 98+%, pure, Thermo Scientific Chemicals
Formic acid, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, 98+%, pure, Thermo Scientific Chemicals
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Plastic bottle
Catalog number FSA147930010
Price (MYR)
754.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
754.00
Add to cart
Chemical Identifiers
CAS6153-56-6
IUPAC Nameoxalic acid dihydrate
Molecular FormulaC2H6O6
InChI KeyGEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILESO.O.OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (ICP-OES)=<5 ppm
Residue after ignition=<0.01 %
Appearance (Form)Crystalline powder
Appearance (Color)White