1-Dodecanesulfonic acid, sodium salt, 99+%
1-Dodecanesulfonic acid, sodium salt, 99+%
1-Dodecanesulfonic acid, sodium salt, 99+%
1-Dodecanesulfonic acid, sodium salt, 99+%
Thermo Scientific Chemicals
1-Dodecanesulfonic acid, sodium salt, 99+%
CAS: 2386-53-0 | C12H25NaO3S | 272.38 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1479000101 gGlass bottle
Catalog number FSA147900010
Price (MYR)
353.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
353.00
Add to cart
Chemical Identifiers
CAS867-56-1
IUPAC Namesodium 2-hydroxypropanoate
Molecular FormulaC3H5NaO3
InChI KeyNGSFWBMYFKHRBD-UHFFFAOYNA-M
SMILES[Na+].CC(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Sodium (Na)12.1 to 12.6 %
pH6.5 to 8.5
Color scale=<50 APHA
Density(20°C) 1.32 to 1.34 g/ml