1-Adamantanecarboxylic acid, 99%, Thermo Scientific Chemicals
1-Adamantanecarboxylic acid, 99%, Thermo Scientific Chemicals
1-Adamantanecarboxylic acid, 99%, Thermo Scientific Chemicals
1-Adamantanecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Adamantanecarboxylic acid, 99%, Thermo Scientific Chemicals
CAS: 828-51-3 | C11H16O2 | 180.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA147880050
Price (MYR)
219.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
219.00
Add to cart
Specifications
Chemical Name or Material1-Adamantanecarboxylic acid
Name Note99%
CAS828-51-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning