Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
CAS: 85715-60-2 | C10H13N2Na3O8 | 358.19 g/mol
Have Questions?
Catalog number FSA147865000
Price (MYR)
696.00
Add to cart
Request bulk or custom format
Price (MYR)
696.00
Add to cart
Chemical Identifiers
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<0.005 %
Calcium (Ca)=<0.005 %
Copper (Cu)=<0.002 %
Lead (Pb)=<0.002 %
Appearance (Color)Orange to red to pink-brown to red-brown
View more
Chelation/Complexation reagent
RUO – Research Use Only