Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Biological buffer, Chelation/Complexation reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA147855000
Price (MYR)
889.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
889.00
Add to cart
Chemical Identifiers
CAS102-49-8
IUPAC Name1-(3,4-dichlorophenyl)methanamine
Molecular FormulaC7H7Cl2N
InChI KeyIXHNFOOSLAWRBQ-UHFFFAOYSA-N
SMILESNCC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.5775 to 1.5795 (20°C, 589 nm)
Appearance (Form)Liquid
Biological buffer, Chelation/Complexation reagent
RUO – Research Use Only