Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Trifluoroacetic anhydride, 99+%, Acros Organics, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number FSA147815000
Price (MYR)
1,541.00
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,541.00
Add to cart
Chemical Identifiers
CAS873-83-6
IUPAC Name6-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H5N3O2
InChI KeyLNDZXOWGUAIUBG-UHFFFAOYSA-N
SMILESNC1=CC(=O)NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Cream to light brown
HPLC>=97.5 %
Acylation reagent
RUO – Research Use Only