Salicylic Acid, 99+%, Thermo Scientific Chemicals
Salicylic Acid, 99+%, Thermo Scientific Chemicals
Salicylic Acid, 99+%, Thermo Scientific Chemicals
Salicylic Acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Salicylic Acid, 99+%, Thermo Scientific Chemicals
CAS: 69-72-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA147700010
Price (MYR)
554.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
554.00
Add to cart
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Phosphate (PO4)=<0.001 %
Titration after Ion exchange>=99.0 %
pH5.2 to 9.2 (5% soln. at 25°C)
Insoluble matter=<0.01 %
Loss on ignition=<0.5 %