Check Order Status
Cyclopentanemethanol, 98%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
Cyclopentanemethanol, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
10 g
Packaging:
Glass bottle
Catalog number
FSA147150100
Price (MYR)
1,117.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,117.00
Add to cart
Specifications
Chemical Name or Material
Cyclopentanemethanol
Name Note
96%
CAS
3637-61-4
Color
Colorless
Density
0.9200g/mL