Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA146750025
Price (MYR)
678.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
678.00
Add to cart
Specifications
Chemical Name or MaterialEthylene glycol
Melting Point2°C to 3°C
Name Note99+%
CAS107-21-1
Health Hazard 1Warning