Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA146750010
Price (MYR)
603.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
603.00
Add to cart
Chemical Identifiers
CAS523-21-7
IUPAC Namedisodium tetraoxocyclohex-1-ene-1,2-bis(olate)
Molecular FormulaC6Na2O6
InChI KeySXWPCWBFJXDMAI-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=97.5 %
Specific absorbance A (1%/1cm)>=450 (at lambda max)
Suitability(for Ba determination) Passes test
Appearance (Color)Grey-green to dark green