1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
CAS: 119-64-2 | C10H12 | 132.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number FSA146730025
Price (MYR)
1,183.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,183.00
Add to cart
Specifications
Chemical Name or Material1,2,3,4-Tetrahydronaphthalene
CAS119-64-2
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0