Safranin O, 95%, pure, high purity biological stain, Thermo Scientific Chemicals
Safranin O, 95%, pure, high purity biological stain, Thermo Scientific Chemicals
Safranin O, 95%, pure, high purity biological stain, Thermo Scientific Chemicals
Safranin O, 95%, pure, high purity biological stain, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Safranin O, 95%, pure, high purity biological stain, Thermo Scientific Chemicals
Safranine O, C20H19ClN4, CAS Number-477-73-6, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride, Basic Red 2, C.I. 50240, 100g, 95%, Brown or Brown-Red to Green, 94% min., 350.84, Pure, Warning, GHS H Statement: Causes serious eye irritation. Causes skin irritation.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA146641000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88-17-5
IUPAC Name2-(trifluoromethyl)aniline
Molecular FormulaC7H6F3N
InChI KeyVBLXCTYLWZJBKA-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4800 to 1.4830 (20°C, 589 nm)
Indicator
RUO – Research Use Only