1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
CAS: 637-87-6 | C6H4ClI | 238.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA146480250
Price (MYR)
391.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
391.00
Add to cart
Chemical Identifiers
CAS79917-90-1
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium chloride
Molecular FormulaC8H15ClN2
InChI KeyFHDQNOXQSTVAIC-UHFFFAOYSA-M
SMILES[Cl-].CCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.0 %
Water=<1.0 % (K.F.)
Appearance (Color)Cream to yellow to orange
Infrared spectrumConforms
Appearance (Form)Powder or crystals or chunks or fused solid