2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
CAS: 135-73-9 | C14H11BrO | 275.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA146350250
Price (MYR)
992.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
992.00
Add to cart
Specifications
Chemical Name or Material2-Bromo-4'-phenylacetophenone
Name Note98%
CAS135-73-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger