1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
Thermo Scientific Chemicals
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA146310050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6000-43-7
SpecificationsSpecification SheetSpecification Sheet
Melting point175°C to 180°C
Water=<1.0 % (K.F.)
Appearance (Color)White to almost white
Infrared spectrumConforms
Anticaking agent0.5 % (SiO2)
View more
Indicator
RUO – Research Use Only