Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number FSA140750010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10043-01-3
IUPAC Namealuminium(3+) potassium disulfate
Molecular FormulaAlKO8S2
InChI KeyGRLPQNLYRHEGIJ-UHFFFAOYSA-J
SMILES[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=15 % (Al) Typical
Appearance (Form)Powder and/or chunks
Total trace metal impurities=<0.001 %
Appearance (Color)White
For analysis
RUO – Research Use Only