DL-Tropic acid, 97%, Thermo Scientific Chemicals
DL-Tropic acid, 97%, Thermo Scientific Chemicals
DL-Tropic acid, 97%, Thermo Scientific Chemicals
DL-Tropic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Tropic acid, 97%, Thermo Scientific Chemicals
CAS: 552-63-6 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA140550250
Price (MYR)
588.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
588.00
Add to cart
Chemical Identifiers
CAS95-57-8
IUPAC Name2-chlorophenol
Molecular FormulaC6H5ClO
InChI KeyISPYQTSUDJAMAB-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to brown
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5580 to 1.5600 (20°C, 589 nm)
GC>=98.0 %