Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
CAS: 101-02-0 | C18H15O3P | 310.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass Bottle
Catalog number FSA140440025
Price (MYR)
807.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
807.00
Add to cart
Chemical Identifiers
CAS7698-05-7
IUPAC Nameproton chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-FTGQXOHASA-N
SMILES[1H+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
D-Enrichment>=99.75 %
Strength0.95 to 1.05 M