2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 527-60-6 | C9H12O | 136.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA140210500
Price (MYR)
879.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
879.00
Add to cart
Chemical Identifiers
CAS1623-08-1
IUPAC Namebis(benzyloxy)phosphinic acid
Molecular FormulaC14H15O4P
InChI KeyHDFFVHSMHLDSLO-UHFFFAOYSA-N
SMILESOP(=O)(OCC1=CC=CC=C1)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point76°C to 80°C
Titration with NaOH>=97.5 %
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)White to light yellow