2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 527-60-6 | C9H12O | 136.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA140210100
Price (MYR)
321.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
321.00
Add to cart
Specifications
Chemical Name or Material2, 4, 6-Trimethylphenol
CAS527-60-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0