Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Catalog number FSA139721000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7790-94-5
IUPAC Namesulfurochloridic acid
Molecular FormulaClHO3S
InChI KeyXTHPWXDJESJLNJ-UHFFFAOYSA-N
SMILESOS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min