Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Catalog number FSA139720250
Price (MYR)
366.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
366.00
Add to cart
Chemical Identifiers
CAS1318-02-1
IUPAC Nameoxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular FormulaAl2O5Si
InChI KeyHNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILESO=[Si]=O.O=[Al]O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (unspecified)Report SiO₂:Al₂O₃ mole ratio, bulk (approx. 12:1)
Assay (unspecified)Report Surface Area