Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
500 g
1 kg
Packaging:
Glass Bottle
Catalog number FSA139720025
Price (MYR)
3,754.00
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
3,754.00
Add to cart
Chemical Identifiers
CAS5423-07-4
IUPAC Namedisodium 3-[(1E)-2-(2,4-dinitrophenyl)diazen-1-yl]-4-hydroxynaphthalene-2,7-disulfonate
Molecular FormulaC16H8N4Na2O11S2
InChI KeyXBLIFEQTVVSTIM-QIKYXUGXSA-L
SMILES[Na+].[Na+].OC1=C2C=CC(=CC2=CC(=C1N=NC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV spectrophotometry>=85 %
Visual transition intervalfrom pH 6.0(bright yellow) to pH 7.2(bright blue)
Appearance (Color)Yellow to orange to brown