Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Catalog number FSA139720010
Price (MYR)
2,389.00
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,389.00
Add to cart
Specifications
Chemical Name or MaterialTrifluoroacetic acid
CAS76-05-1
Melting Point-15.0°C
Boiling Point72.0°C
Health Hazard 1Danger