Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
CAS: 867-13-0 | C8H17O5P | 224.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA139705000
Price (MYR)
1,724.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,724.00
Add to cart
Chemical Identifiers
CAS199006-54-7
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-methylphenyl)propanoic acid
Molecular FormulaC25H23NO4
InChI KeyUXLHLZHGQPDMJQ-QHCPKHFHSA-N
SMILESCC1=CC=C(C[C@H](NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥94.0%
Water Content (Karl Fischer Titration)≤1.0%
Melting Point (clear melt)165.0-175.0?C
View more
for C-C Bond Formation (Olefination)
RUO – Research Use Only