2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA139250250
Price (MYR)
445.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
445.00
Add to cart
Chemical Identifiers
CAS61272-71-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=94.0 %
Appearance (Color)White to yellow to light brown
Infrared spectrumConforms
Melting point174°C to 178°C