Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
CAS: 102-76-1 | C9H14O6 | 218.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA139225000
Price (MYR)
441.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
441.00
Add to cart
Chemical Identifiers
CAS393-09-9
IUPAC Name4-fluoro-1-nitro-2-(trifluoromethyl)benzene
Molecular FormulaC7H3F4NO2
InChI KeyWMQOSURXFLBTPC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4580-1.4620 @ 20?C
Appearance (Color)Clear yellow
Assay (GC)≥98.0%