Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
CAS: 102-76-1 | C9H14O6 | 218.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
Glass bottle
Catalog number FSA139220025
Price (MYR)
1,087.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,087.00
Add to cart
Specifications
Chemical Name or MaterialTriacetin
CAS102-76-1
Melting Point3.0°C
ColorColorless
Density1.1550g/mL