Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
CAS: 102-76-1 | C9H14O6 | 218.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA139220010
Price (MYR)
615.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
615.00
Add to cart
Chemical Identifiers
CAS59279-60-6
IUPAC Name1,5-dimethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}pentanedioate
Molecular FormulaC12H21NO6
InChI KeyQNSPKWUAZQIIGZ-QMMMGPOBSA-N
SMILESCOC(=O)CC[C@H](NC(=O)OC(C)(C)C)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%