o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
CAS: 95-53-4 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass bottle
Catalog number FSA139080025
Price (MYR)
1,200.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,200.00
Add to cart
Specifications
Chemical Name or Materialo-Toluidine
Name Note99%
CAS95-53-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Toxic if inhaled.
Suspected of causing genetic defects if inhaled.
May cause cancer by inhalat