p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 98-59-9 | C7H7ClO2S | 190.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA139030020
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS86639-52-3
IUPAC Name(19S)-10,19-diethyl-7,19-dihydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Molecular FormulaC22H20N2O5
InChI KeyFJHBVJOVLFPMQE-QFIPXVFZSA-N
SMILESCCC1=C2C=C(O)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Off-white to yellow
Appearance (Form)Powder
HPLC>=97.5 %
Water=<5 %