p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
CAS: 207801-20-5 | C7H7NaO2S | 178.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA139011000
Price (MYR)
494.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
494.00
Add to cart
Chemical Identifiers
CAS311-28-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Powder or crystals or flakes
Infrared spectrumConforms
Melting point141°C to 150°C
Titration Argentometric>=97.5 to 102.5 %