Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, Thiocarbamide, CAS # 62-56-6, is an analogue of urea with a single sulfur-substituted oxygen.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
1 kg
Packaging:
View all
Plastic drum
Catalog number FSA138910050
Price (MYR)
1,947.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,947.00
Add to cart
Chemical Identifiers
CAS121-92-6
IUPAC Name3-nitrobenzoic acid
Molecular FormulaC7H5NO4
InChI KeyAFPHTEQTJZKQAQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point139.5-145.5?C
Appearance (Color)White to cream or pale yellow
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
View more
General description
• Thiourea is a derivative of the biological compound, urea
• Thiourea is highly toxic when it decomposes and acts as a carcinogen
Applications
• Thiourea can be used as a denaturant in lysis buffers and similar sample buffers that require solubilization of proteins
• Thiourea can act as an antioxidant and chromophore
• Thiourea can be used in the study of cellular redox, autophagy, and aging
RUO – Research Use Only
General References:
  1. Wang, Y.; Jin, X.; He, L.; Zhang, W. Inhibitory effect of thiourea on biological nitrification process and its eliminating method. Water Sci Technol. 2017 75(12), 2900-2907.
  2. Park, S.; Lee, H.; Chae, J.B.; Kim, C. A Thiourea-Containing Fluorescent Chemosensor for Detecting Ga3. J Fluoresc. 2020 30(6), 1457-1462.