Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, Thiocarbamide, CAS # 62-56-6, is an analogue of urea with a single sulfur-substituted oxygen.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA138910010
Price (MYR)
498.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
498.00
Add to cart
Chemical Identifiers
CAS102-49-8
IUPAC Name1-(3,4-dichlorophenyl)methanamine
Molecular FormulaC7H7Cl2N
InChI KeyIXHNFOOSLAWRBQ-UHFFFAOYSA-N
SMILESNCC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.5760-1.5810 @ 20?C
General description
• Thiourea is a derivative of the biological compound, urea
• Thiourea is highly toxic when it decomposes and acts as a carcinogen
Applications
• Thiourea can be used as a denaturant in lysis buffers and similar sample buffers that require solubilization of proteins
• Thiourea can act as an antioxidant and chromophore
• Thiourea can be used in the study of cellular redox, autophagy, and aging
RUO – Research Use Only
General References:
  1. Wang, Y.; Jin, X.; He, L.; Zhang, W. Inhibitory effect of thiourea on biological nitrification process and its eliminating method. Water Sci Technol. 2017 75(12), 2900-2907.
  2. Park, S.; Lee, H.; Chae, J.B.; Kim, C. A Thiourea-Containing Fluorescent Chemosensor for Detecting Ga3. J Fluoresc. 2020 30(6), 1457-1462.