Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA138891000
Price (MYR)
432.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
432.00
Add to cart
Chemical Identifiers
CAS123-11-5
IUPAC Name4-methoxybenzaldehyde
Molecular FormulaC8H8O2
InChI KeyZRSNZINYAWTAHE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Liquid
Relative density(D20/4°C) 1.120 to 1.124
Appearance (Color)Clear colorless to yellow