2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
CAS: 79-42-5 | C3H6O2S | 106.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Catalog number FSA138791000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
Assay (Titration)70.0 to 74.0 % (w/w)