Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
CAS: 507-09-5 | C2H4OS | 76.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA138785000
Price (MYR)
1,199.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,199.00
Add to cart
Chemical Identifiers
CAS4433-40-3
IUPAC Name5-(hydroxymethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC5H6N2O3
InChI KeyJDBGXEHEIRGOBU-UHFFFAOYSA-N
SMILESOCC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)(Based on Carbon content): >97.5 to <102.5%
Elemental AnalysisNitrogen: 19.21-20.19% (Theory: 19.70%)
Appearance (Color)White to cream or grey
FormPowder
Elemental AnalysisCarbon: 41.16-43.28% (Theory: 42.22%)